Learn More
Medchemexpress LLC 1H-Benzimidazole-2-methanamine, N-[[3-ethoxy-4-(2-thienylmethoxy)phenyl]methyl]- | 878949-21-4 | 99.31% | 393.50 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1716165MG
Encompass_Preferred
DCEM1 binds to heat shock protein 60 (HSP60) and inhibits its interaction with ClpP, which blocks the mitochondrial unfolded protein response. It also inhibits β-catenin expression and ATP production in PC-3 and TKO cells. DCEM1 can be used in prostate cancer research.
  • Binds to heat shock protein 60 (HSP60)
  • Inhibits the interaction of HSP60 with ClpP
  • Blocks the mitochondrial unfolded protein response
  • Inhibits β-catenin expression
  • Inhibits ATP production in PC-3 and TKO cells
  • Can be used in prostate cancer research
Catalog No. 50-003-86461