Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-Benzimidazole-5-carboxylic acid, 2-[[[4-[[[(hexyloxy)carbonyl]amino]iminomethyl]phenyl]amino]methyl]-1-methyl-, ethyl ester | 1408238-36-7 | 479.57 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYW004366

Encompass_Preferred

Dabigatran impurity 75 is an off-white to yellow solid with the molecular formula C26H33N5O4 and a molecular weight of 479.57. It is characterized by its 1H NMR spectrum being consistent with its structure and has a purity of ≥97.0%. This product is for research use only and has not been fully validated for medical applications.

  • Off-white to yellow solid appearance
  • Purity of ≥97.0% (NMR)
  • Molecular formula C26H33N5O4
  • Molecular weight of 479.57

Catalog No. 50-004-49386


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.