Learn More
Medchemexpress LLC 1H-Benzimidazole, 6-bromo-4-fluoro-2-methyl-1-(1-methylethyl)- | 1231930-33-8 | >99.4% | C11H12BrFN2 | 100 G

Supplier: Medchemexpress LLC HY78066100G
Abemaciclib Impurity 1 is a high-purity chemical compound from MedChemExpress, available as an off-white to yellow solid. This impurity features the molecular formula C11H12BrFN2 and a molecular weight of 271.13. It is commonly used in chemical synthesis and research applications.
- Purity of >99.4%
- Storage as powder: -20°C for 3 years, or 4°C for 2 years
- Storage in solvent: -80°C for 6 months, or -20°C for 1 month
- Complies with specified quality standards
- Distinctive off-white to yellow solid appearance
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.