Learn More
Medchemexpress LLC 1H-Benzimidazole, 6-methoxy-2-[(S)-[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfinyl]-, sodium salt (1:1) | 161796-78-7 | MFCD00870347 | 99.4% | 367.40 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY170235MG
Encompass_Preferred
Esomeprazole sodium is a potent and orally active proton pump inhibitor. It reduces acid secretion by inhibiting the H+, K+-ATPase in gastric parietal cells. Additionally, it acts as an exosome inhibitor by blocking exosome release through the inhibition of V-H+-ATPases. It has potential for research related to symptomatic gastroesophageal reflux disease and for suppressing the growth of triple-negative breast cancer cells.
  • Potent and orally active proton pump inhibitor
  • Reduces acid secretion
  • Acts as an exosome inhibitor
  • Potential for symptomatic gastroesophageal reflux disease research
  • For research use only
Catalog No. 50-003-84294