Learn More
Medchemexpress LLC 1H-Imidazole, 1-[(4-methylphenyl)sulfonyl]- | 2232-08-8 | 222.27 | 50 G

Supplier: Medchemexpress LLC HYW00196550G
1-Tosylimidazole is a biochemical reagent suitable for life science research. It can be utilized as a biological material or an organic compound. This product is for research use only.
- High purity (99.75%)
- White to off-white solid appearance
- Molecular formula C10H10N2O2S
- Soluble in DMSO (100 mg/mL)
- Stable for extended storage as powder at -20°C (3 years) or 4°C (2 years)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.