Learn More
Medchemexpress LLC 1H-Imidazole-1-acetonitrile, 2-[[2-(2,3-dihydro-1,4-benzodioxin-6-yl)-2-oxoethyl]thio]- | 2418577-51-0 | 99.6% | 315.35 | 25 MG

Supplier: Medchemexpress LLC 25MGHY146985
Cathepsin X-IN-1 (compound 25) is a potent Cathepsin X inhibitor (IC50 of 7.13 μM). It decreases PC-3 cell migration with low cytotoxicity.
- Potent Cathepsin X inhibitor.
- IC50 of 7.13 μM against Cathepsin X.
- Decreases PC-3 cell migration.
- Exhibits low cytotoxicity.
- For research use only.
- Purity: 99.62%.
- Molecular weight: 315.35.
- Formula: C15H13N3O3S.
- CAS no.: 2418577-51-0.
- Appearance: Solid, white to off-white.
- Storage: 4°C (protected from light); in solvent: -80°C (6 months), -20°C (1 month), protected from light.
- Soluble in DMSO (≥ 100 mg/mL); soluble in H2O (4.85 mg/mL) with ultrasonic, warming, and adjusting pH to 3 with HCl and heating to 60°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.