Learn More
Medchemexpress LLC 1H-Imidazole-2-carbaldehyde | 10111-08-7 | 96.09 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW007321
Encompass_Preferred
1H-Imidazole-2-carbaldehyde is a biochemical reagent used in life science research. It functions as a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor, with applications in suppressing type-2 diabetes. Additionally, it is utilized in the preparation of tridentate Schiff-base carboxylate-containing ligands and studies involving imidazole-directed allylation of aldimines.
  • Biochemical reagent for life science research.
  • Functions as a novel protein tyrosine phosphatase 1B (PTP1B) inhibitor.
  • Applied to suppress type-2 diabetes.
  • Used in the preparation of tridentate Schiff-base carboxylate-containing ligands.
  • Involved in the study of imidazole-directed allylation of aldimines.
Catalog No. 50-004-48205