Learn More
Medchemexpress LLC 1H-imidazole-4-carboxaldehyde, 1-(triphenylmethyl)- | 33016-47-6 | 99.1% | C23H18N2O | 10 G
SDP
Supplier:  Medchemexpress LLC HY7704910G
Encompass_Preferred
1-Trityl-1H-imidazole-4-carbaldehyde is a biochemical reagent used as a biological material or organic compound for life science-related research. It is for research use only and not intended for medical applications.
  • Used as a biochemical reagent
  • Applicable in life science-related research
  • Used in the synthesis of histamine and girolline (in vitro)
  • Appears as a white to off-white solid
Catalog No. 50-004-34945