Learn More
Medchemexpress LLC 1h-indazole-4-carboxamide, n-[(1,2-dihydro-1,6-dimethyl-2-oxo-4-propyl-3-pyridinyl)methyl]- | 1433200-49-7 | 99.9% | 1 ML

Supplier: Medchemexpress LLC HY1284510MM/1ML
UNC2400 is a close analog of UNC1999, serving as a negative control for cell-based studies with >1,000-fold lower potency than UNC1999. This solid, white to off-white compound has a molecular weight of 597.79 and a chemical formula of C35H47N7O2.
- Used as a negative control for cell-based studies.
- Purity of 99.92%.
- Molecular weight is 597.79.
- Chemical formula is C35H47N7O2.
- Appearance as a white to off-white solid.
- Histone methyltransferase inhibitor.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.