Learn More
Medchemexpress LLC 1H-Inden-1-one, 2,3-dihydro-5,6-dimethoxy-2-(4-piperidinylmethyl)-, hydrochloride (1:1) | 120013-39-0 | 325.84 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW003576
Encompass_Preferred
ACHE-IN-38 hydrochloride is an analytical standard and research compound that inhibits the metabolic breakdown of acetylcholine (ACh) by acetylcholinesterase (AChE). This action helps alleviate memory deficits in Alzheimer's Disease patients by potentiating cholinergic transmission.
  • Inhibits metabolic breakdown of acetylcholine (ACh)
  • Targets acetylcholinesterase (AChE) enzyme
  • Potentiates cholinergic transmission
  • May help alleviate memory deficits in Alzheimer's Disease patients
  • Intended for research and analytical applications
  • Suitable for qualitative, quantitative, and methodological research experiments (e.g., HPLC, GC, MS)
Catalog No. 50-004-48191