Learn More
Medchemexpress LLC 1H-Indol-5-amine (5-Aminoindole) | 5192-03-0 | 132.17 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0075095G
Encompass_Preferred
1H-Indol-5-amine (5-Aminoindole) is an organic compound featuring an indole ring structure, classified as an indole derivative. It serves as a crucial intermediate for synthesizing other active compounds.
  • Undergoes a three-component reaction with aromatic aldehydes and tert-butyl 2,4-dioxopiperidine-1-carboxylate to yield 11-aryl-3H-indolo[5,4-b]naphthyridine derivatives.
  • Used as a ligand for preparing hydrophobic charge induction chromatography medium for protein adsorption and elution, with potential for protein purification.
Catalog No. 50-004-47733