Learn More
Medchemexpress LLC 1H-indole-1-carboxylic acid, 5-bromo-, 1,1-dimethylethyl ester | 182344-70-3 | MFCD02090067 | 100.0% | 296.16 g·mol⁻¹ | C13H14BrNO2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW02159710G
Encompass_Preferred
N-Boc-5-bromoindole is a tert-butyl carbamate (Boc) protected 5-bromoindole used as an intermediate in organic synthesis. It is commonly employed in Buchwald-Hartwig amination workflows to prepare protected 5-aminoindole derivatives and other functionalized indoles. The compound is supplied as a high-purity solid intended for research and synthetic applications.
  • Boc-protected indole suitable for selective N-functionalization.
  • High purity for reliable synthetic performance.
  • Solid, white to off-white appearance for easy handling.
  • Stable under recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years).
  • Available in small laboratory package sizes for bench synthesis.
Catalog No. 50-003-00961