Learn More
Medchemexpress LLC 1H-Indole-1-carboxylic acid, octahydro-4-hydroxy-4-[2-(3-methylphenyl)ethynyl]-, methyl ester, (3aR,4S,7aR)- | 543906-09-8 | C19H23NO3 | 1 G
SDP
Supplier:  Medchemexpress LLC HY152571G
Encompass_Preferred
Mavoglurant is a potent, selective, non-competitive, and orally active mGluR5 antagonist. It is suitable for research into Fragile X syndrome (FXS) and L-dopa induced dyskinesias in Parkinson's disease. This compound also functions as a click chemistry reagent due to its alkyne group, enabling participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with azide-containing molecules.
  • Potent, selective, non-competitive, and orally active mGluR5 antagonist
  • IC50 of 30 nM for mGluR5
  • Exhibits over 300-fold selectivity for mGluR5 over 238 other targets
  • Suitable for research into Fragile X syndrome (FXS) and L-dopa induced dyskinesias in Parkinson's disease
  • Functions as a click chemistry reagent
  • Contains an alkyne group enabling participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
Catalog No. 50-003-73001