Learn More
Medchemexpress LLC 1H-Indole-2-carboxamide, 4-fluoro-7-methyl-N-[(1R,3S)-3-[(3S)-3-[methyl(methylsulfonyl)... | 2697176-16-0 | 99.00% | 450.57 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13962650MG
Encompass_Preferred
EPZ-719 is a selective, orally effective SETD2 inhibitor with an IC50 value of 0.005 μM. It demonstrates anticancer activity. This product is for research use only and is not intended for patient use. In vitro studies show EPZ-719 (after 14 days) can inhibit cell proliferation in KMS34 and KMS11 cells with IC50 values of 0.025 μM and 0.211 μM, respectively.
  • Selective, orally effective SETD2 inhibitor
  • IC50 value of 0.005 μM
  • Demonstrates anticancer activity
  • Inhibits cell proliferation in KMS34 and KMS11 cells
  • Intended for research use only
Catalog No. 50-003-50451