Learn More
Medchemexpress LLC 1H-Indole, 3-(2-chloro-4-pyrimidinyl)-1-methyl- | 1032452-86-0 | 99.5% | C13H10ClN3 | 100 G

Supplier: Medchemexpress LLC HY78866100G
3-(2-Chloropyrimidin-4-yl)-1-methylindole (MedChemExpress HY-78866) is a high-purity chemical compound, appearing as an off-white to light yellow solid. With a molecular weight of 243.70 and a purity of 99.5%, it is suitable for various research applications.
- Appearance: Off-white to light yellow solid
- Purity: 99.5%
- Powder storage stability: 3 years at -20°C, 2 years at 4°C
- In solvent storage stability: 6 months at -80°C, 1 month at -20°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.