Learn More
Medchemexpress LLC 1H-Indole-3-acetic acid, α-(2-oxo-2-phenylethyl)- | 6266-66-6 | 99.6% | 293.32 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11273010MM/1ML
Encompass_Preferred
PEO-IAA is an indole-3-acetic acid (IAA) antagonist. It acts as an auxin antagonist that binds to transport inhibitor response 1/auxin signaling F-box proteins (TIR1/AFBs).
  • Shows potent anti-auxin activity in auxin-responsive gene expression and in the cell division and elongation pathway that is mediated via SCFTIR1/AFBs.
  • Suppresses the expression of an auxin-responsive ZmSAUR2 gene and gravitropic curvature.
  • Blocks the auxin response in Arabidopsis, rice, moss and maize.
Catalog No. 50-004-37876