Learn More
Medchemexpress LLC 1H-Indole, 5-bromo- | 10075-50-0 | 99.97% | 196.05 | 10 G
SDP
Supplier:  Medchemexpress LLC HY3023610G
Encompass_Preferred
5-Bromoindole is an antibacterial agent with a Minimum Inhibitory Concentration (MIC) of 200 μg/mL. It functions by inhibiting biofilm and curli formation, and by reducing both swarming and swimming motility. This agent can be utilized for research pertaining to enterohemorrhagic Escherichia coli O157:H7 infection.
  • Antibacterial agent with a MIC of 200 μg/mL.
  • Inhibits biofilm and curli formation.
  • Reduces swarming motility.
  • Reduces swimming motility.
  • Can be used for the research of enterohemorrhagic Escherichia coli O157:H7 infection.
Catalog No. 50-004-04376