Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-indole-6-carboxylic acid | 1670-82-2 | MFCD00210441 | 99.2% | 161.16 g·mol⁻¹ | C9H7NO2 | 10 G
SDP

Supplier:  Medchemexpress LLC HYW01542010G

Encompass_Preferred

1H-indole-6-carboxylic acid is an organic compound used as a biochemical reagent in life-science and medicinal chemistry research. It functions as a building block and intermediate for synthesis of indole derivatives and is employed in analytical and preparative applications.

  • High purity (99.23%) suitable for research applications.
  • Molecular formula C9H7NO2 and molecular weight 161.16 g·mol⁻¹.
  • Available in small laboratory quantities, including 10 g packaging.
  • Suitable for organic synthesis and medicinal chemistry workflows.
  • Offered as a stable solid for convenient handling and storage.

Catalog No. 50-003-02952


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.