Learn More
Medchemexpress LLC 1H-indole-6-carboxylic acid | 1670-82-2 | MFCD00210441 | 99.2% | 161.16 g/mol | C9H7NO2 | 5 G

Supplier: Medchemexpress LLC HYW0154205G
1H-Indole-6-carboxylic acid (CAS 1670-82-2) is an indole-derived biochemical reagent (C9H7NO2, MW 161.16 g/mol) supplied for research use. It is provided as a high-purity organic compound for life-science and chemical research applications.
- High purity: 99.23%.
- Intended for research use only.
- Suitable as an organic reagent for life-science research.
- Powder is stable with storage at -20°C (3 years) and 4°C (2 years).
- Available in multiple pack sizes: 5 g, 10 g, 25 g, 50 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.