Learn More
Medchemexpress LLC 1H-Indole-7-carbaldehyde | 1074-88-0 | MFCD01318152 | 145.16 | 10 G

Supplier: Medchemexpress LLC HYW00801310G
1H-Indole-7-carbaldehyde is a biochemical reagent suitable for life science related research. It is intended for research use only and not for patient use or medical applications. This compound is supplied as an off-white to yellow solid.
- Can be used as a biological material or organic compound
- Molecular weight of 145.16
- Molecular formula C9H7NO
- Store at 4°C, under nitrogen
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.