Learn More
Medchemexpress LLC 1H-isoindole-1,3(2H)-dione, 2-(2,6-dioxo-3-piperidinyl)-5-iodo | 2509137-01-1 | MFCD31735039 | 98.0% | 384.13 | C13H9IN2O4 | 500 MG

Supplier: Medchemexpress LLC HYW586331500MG
Thalidomide-iodo is an iodinated derivative of thalidomide used as an E3 ligase (cereblon) activator for research applications. It is intended as a building block and synthetic intermediate for further derivatization by substitution of the iodine atom.
- E3 ligase activator (cereblon ligand) for targeted protein degradation research.
- Suitable as a synthetic intermediate for iodide substitution and further derivatization.
- High purity: 98.0% (NMR).
- Molecular formula C13H9IN2O4; molecular weight 384.13.
- CAS number 2509137-01-1.
- Appearance: gray to dark gray solid.
- Storage: 4°C, protect from light; in solvent: -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.