Learn More
Medchemexpress LLC 1h-Isoindole-1,3(2h)-dione, 5,6-bis[(4-fluorophenyl)amino]- | 145915-60-2 | 98.9% | 365.33 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10860050MG
Encompass_Preferred
CGP-53353 (DAPH-7) is a potent PKC inhibitor with IC50s of 0.41 μM and 3.8 μM for PKCβII and PKCβI, respectively. It inhibits glucose-induced cell proliferation and DNA synthesis in AoSMC and A10 cells and can be used for researching atherosclerosis in diabetic patients.
  • Potent PKC inhibitor
  • Inhibits glucose-induced cell proliferation and DNA synthesis in AoSMC and A10 cells
  • Can be used for researching atherosclerosis of diabetic patients
Catalog No. 50-003-44764