Learn More
Medchemexpress LLC 1H-Isoindole-1,3(2H)-dione, 5-amino-2-[2,6-bis(1-methylethyl)phenyl]- | 100823-03-8 | 99.55% | 50 MG

Supplier: Medchemexpress LLC HY12947850MG
TC11 is a MCL1 degrader and Caspase-9 and CDK1 activator. It is a phenylacetylamide derivative structurally related to immunomodulatory active molecules. TC11 induces the degradation of MCL1, leading to apoptotic death during prolonged mitotic arrest.
- MCL1 degrader
- Caspase-9 and CDK1 activator
- Induces apoptotic death
- Phenylacetylamide derivative
- Purity: 99.55%
- Soluble in DMSO (50 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.