Learn More
Medchemexpress LLC 1H-Pyrano[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione, 4,7-diethyl-4,10-dihydroxy-, (S)- | 119577-28-5 | 392.40 | 100 MG
Supplier: Medchemexpress LLC HYN2063100MG
12-Ethyl-9-hydroxycamptothecin is a derivative of Camptothecin. Camptothecin (CPT), a kind of alkaloid, is a DNA topoisomerase I (Topo I) inhibitor with an IC50 of 679 nM.
- Derivative of Camptothecin
- Alkaloid
- DNA topoisomerase I (Topo I) inhibitor
- IC50 of 679 nM
- For research use only