Learn More
Medchemexpress LLC 1H-Pyrano[3,4-f]indolizine-3,6,10(4H)-trione, 4-ethyl-7,8-dihydro-4-hydroxy- | 102978-40-5 | 100.0% | 263.25 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY42487A
Encompass_Preferred
(rac)-Exatecan Intermediate 1 is an isomer of Exatecan Intermediate 1 and an intermediate of Exatecan. Exatecan is a camptothecin-based anticancer agent that inhibits tumor growth by interfering with the proliferation and division of tumor cells through interaction with DNA. It is primarily used in research for various cancers, including ovarian, lung, and breast cancer.
  • High purity (99.97%)
  • Targets camptothecins
  • Solid appearance
  • White to light yellow color
  • Soluble in DMSO (100 mg/mL)
Catalog No. 50-004-02429