Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-Pyrano[3,4-f]indolizine-3,6,10(4H)-trione, 4-ethyl-7,8-dihydro-4-hydroxy- | 102978-40-5 | 99.97% | 263.25 | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHY42487A

Encompass_Preferred

(rac)-Exatecan Intermediate 1 is an isomer of Exatecan Intermediate 1 and an intermediate of Exatecan, a camptothecin-based anticancer agent. It inhibits tumor growth by interfering with the proliferation and division of tumor cells by interacting with DNA. This product is primarily used in research into a variety of cancers including ovarian, lung, and breast cancer.

  • Appearance: Solid
  • Color: White to light yellow
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage for powder: -20°C for 3 years, 4°C for 2 years.
  • Storage in solvent: -80°C for 6 months, -20°C for 1 month.

Catalog No. 50-004-02469


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.