Learn More
Medchemexpress LLC 1H-pyrazole-1-carboxylic acid, 3-amino-5-cyclobutyl-, 1,1-dimethylethyl ester | 1026682-51-8 | 237.30 | C12H19N3O2 | 5 MG

Supplier: Medchemexpress LLC HY1074485MG
1H-Pyrazole-1-carboxylic acid, 3-amino-5-cyclobutyl-, 1,1-dimethylethyl ester is a small-molecule drug intermediate (Protac CDK9-binding moiety 1) used in the synthesis of PROTACs and other bioactive compounds. Supplied in milligram quantities for research use only; not for human or clinical use.
- Used as a building block for PROTAC and small-molecule synthesis.
- CAS number 1026682-51-8 for unambiguous identification.
- Molecular formula C12H19N3O2 and molecular weight 237.30.
- Available in small milligram pack sizes suitable for research applications.
- Powder storage: -20 °C; in solvent: -80 °C (6 months) and -20 °C (1 month).
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.