Learn More
Medchemexpress LLC 1H-pyrazole-1-carboxylic acid, 3-amino-5-cyclobutyl-, 1,1-dimethylethyl ester | 1026682-51-8 | 98.1% | 237.30 g/mol | C12H19N3O2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10744850MG
Encompass_Preferred
1H-pyrazole-1-carboxylic acid, 3-amino-5-cyclobutyl-, 1,1-dimethylethyl ester is a small-molecule chemical intermediate used primarily in medicinal chemistry for the synthesis of PROTACs and other bioactive compounds. It is supplied as a high-purity solid suitable for research-scale synthesis and analytical characterization.
  • High purity (98.1%) suitable for synthesis and characterization
  • Molecular weight 237.30 g/mol
  • Solid form stable under recommended storage conditions
  • Useful as a PROTAC CDK9-binding moiety intermediate
  • Available in small pack sizes tailored for medicinal chemistry workflows
Catalog No. 50-002-40773