Learn More
Medchemexpress LLC 1H-Pyrazole, 3-methyl- | 1453-58-3 | 99.5% | C4H6N2 | 25G

Supplier: Medchemexpress LLC 25GHY66054
3-Methylpyrazole is utilized as a nitrification inhibitor in soil and has been shown to inhibit the metabolism of MAM by rat liver microsomes in vitro.
- Utilized as a nitrification inhibitor in soil
- Inhibits the metabolism of MAM by rat liver microsomes
- Appears as a light yellow to reddish brown solid below 36.5°C and liquid above 36.5°C
- Store pure form at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
- Soluble in H2O at 25 mg/mL (304.48 mM)
- Soluble in PBS at 50 mg/mL (608.97 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.