Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-pyrazole-4-carboxylic acid, ethyl ester | 37622-90-5 | MFCD00010844 | 99.8% | 140.14 g/mol | C6H8N2O2 | 100 G
SDP

Supplier:  Medchemexpress LLC HY100040100G

Encompass_Preferred

Ethyl 1H-pyrazole-4-carboxylate (synonym: 4-ethoxycarbonylpyrazole) is an organic intermediate and biochemical reagent used in life-science research and organic synthesis. It is employed to prepare isoxazole-4-carboxylic acid derivatives and isoxazole-3,5-dicarboxamides, and serves as a versatile building block for small-molecule synthesis. Intended for research use only.

  • High purity (99.77%).
  • Molecular weight 140.14 g/mol.
  • CAS number 37622-90-5 for unambiguous identification.
  • Useful intermediate for isoxazole derivative synthesis.
  • Available in laboratory pack sizes for convenient handling.

Catalog No. 50-002-15184


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.