Learn More
Medchemexpress LLC 1H-pyrazole,5-(4-chlorophenyl)-1-(2-methoxyphenyl)-3-(tetrahydro-2,2,6,6-tetramethyl-2H-pyran-4-yl)- | 1567335-29-8 | 98.9% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11937310MM 1ML
Encompass_Preferred
Cav 2.2 blocker 1 is a potent N-type calcium channel (CaV2.2) blocker (compound 9) used in preclinical research on pain. It is supplied as a 10 mM solution in DMSO, with reported in vitro potency (IC50 ≈ 1 nM), a reported purity of 98.85%, molecular weight 424.96, and chemical formula C25H29ClN2O2.
  • Supplied as 10 mM solution in DMSO for immediate experimental use.
  • Reported IC50 ≈ 1 nM against N-type (CaV2.2) calcium channels.
  • High reported purity supports reliable experimental results.
  • Off-white to light yellow solid appearance when isolated.
  • Product documentation includes datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-33902