Learn More
Medchemexpress LLC 1H-Pyrido[3,4-b] indole, 2,3,4,9-tetrahydro- | 16502-01-5 | 99.4% | 172.23 | 5 G
SDP
Supplier:  Medchemexpress LLC HY206965G
Encompass_Preferred
Tetrahydro-β-carboline (Tryptoline) is a metabolite of tryptamine and a competitive serotonin reuptake inhibitor with a Ki value of 6.1 μM. It is for research use only and not sold to patients. It increases serotonin levels in the brain of rats.
  • Metabolite of tryptamine.
  • Competitive serotonin reuptake inhibitor.
  • Increases serotonin levels in the brain (demonstrated in rats).
  • Available in solid form and as a 10 mM solution in DMSO.
  • For research use only.
  • Serotonin: 6.1 μM (Ki).
Catalog No. 50-004-00546