Learn More
Medchemexpress LLC 1H-pyrido[3,4-b]indole-3-carboxylic acid, 1-(2,4-dichlorophenyl)-2,3,4,9-tetrahydro-, (1R,3S)- | 1679333-73-3 | 98.5% | 361.22 g/mol | C18H14Cl2N2O2 | 10MM 1ML

Supplier: Medchemexpress LLC HY12356110MM 1ML
MMV008138 is a small-molecule IspD inhibitor used in antimalarial research and biochemical screening. The compound is provided in both solid form and a ready-to-use 10 mM solution in DMSO, with confirmed molecular formula C18H14Cl2N2O2, molecular weight 361.22 g/mol, and high purity suitable for assay work. Certificate of analysis and product datasheet are available for quality verification.
- Species-selective IspD inhibitor for antimalarial studies.
- High purity suitable for biochemical and cellular assays.
- Available as solid or 10 mM solution in DMSO for convenience.
- Molecular weight 361.22 g/mol and formula C18H14Cl2N2O2.
- Certificate of analysis available for batch verification.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.