Learn More
Medchemexpress LLC 1H-pyrrole-1-butanoic acid, 2,5-dihydro-2,5-dioxo-, 2,5-dioxo-1-pyrrolidinyl ester | 80307-12-6 | MFCD00036817 | 99.7% | 280.23 g·mol⁻1 | C12H12N2O6 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW04199410G
Encompass_Preferred
GMBS is a heterobifunctional cross-linker used to covalently link amine and thiol groups, commonly applied in mapping protein-protein interfaces and structural studies such as chemical cross-linking coupled with mass spectrometry (CXMS).
  • reactive toward primary amines and thiol groups
  • suitable for cross-linking and structural mapping workflows
  • high purity appropriate for analytical applications
  • available in multiple pack sizes, including 10 g
Catalog No. 50-002-08823