Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1H-Pyrrole-1-undecanoic acid, 2,5-dihydro-2,5-dioxo-, 2,5-dioxo-1-pyrrolidinyl ester | 87981-04-2 | 98.0% | 378.42 | 50 MG
SDP

Supplier:  Medchemexpress LLC HYW00903750MG

Encompass_Preferred

Maleimide-C10-NHS ester is an alkyl/ether-based PROTAC linker that can be utilized in the synthesis of PROTACs. PROTACs incorporate two distinct ligands connected by a linker, one targeting an E3 ubiquitin ligase and the other for the protein of interest, thereby employing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • Purity: 98.0%
  • Molecular weight: 378.42
  • Formula: C19H26N2O6
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder at -20°C for 3 years; in solvent at -80°C for 6 months, or -20°C for 1 month
  • Solubility: ≥ 100 mg/mL in DMSO (264.26 mM)

Catalog No. 50-003-66895


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.