Learn More
Medchemexpress LLC 1H-Pyrrole-2,5-dione, 1-(3-aminopropyl)-, hydrochloride (1:1) | 1257852-74-6 | 98.0% | 190.63 | 250 MG

Supplier: Medchemexpress LLC 250MGHY43702A
1-(3-Aminopropyl)-1H-pyrrole-2,5-dione hydrochloride is a chemical compound with a molecular weight of 190.63 and a purity of 98.0%. It is supplied as a white to off-white solid and is recommended for various laboratory applications requiring a high-purity chemical reagent.
- Supplied as a white to off-white solid
- Molecular formula: C7H11CIN2O2
- Storage conditions for powder: -20°C for 3 years, 4°C for 2 years
- Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month
- Conforms to specified analytical data
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.