Learn More
Medchemexpress LLC 1H-Pyrrole-2,5-dione, 1-(3-aminopropyl)-, hydrochloride (1:1) | 1257852-74-6 | MFCD11519188 | 98.0% | 190.63 | 50 MG

Supplier: Medchemexpress LLC HY43702A50MG
This product is 1H-Pyrrole-2,5-dione, 1-(3-aminopropyl)-, hydrochloride (1:1), a biochemical reagent for research use only. It appears as a white to off-white solid.
- Not fully validated for medical applications; for research use only.
- White to off-white solid appearance.
- Molecular weight: 190.63.
- Purity: 98.0%.
- Storage conditions for powder: -20°C for 3 years, 4°C for 2 years.
- Storage conditions in solvent: -80°C for 6 months, -20°C for 1 month.
- Shipping: Room temperature.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.