Learn More
Medchemexpress LLC 1H-pyrrole-2,5-dione, 1-[4-[2-(4-methylphenyl)-1,2-diphenylethenyl]phenyl]- | 1245606-71-6 | 98.9% | C31H23NO2 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14321850MG
Encompass_Preferred
TPE-MI (Tetraphenylethene maleimide) is a thiol probe utilized for measuring unfolded protein load and proteostasis in cells. It has an excitation wavelength of 350 nm and an emission wavelength of 470 nm. This probe can identify proteostasis imbalances in induced pluripotent stem cell models of Huntington disease and detect protein damage in research involving the malaria parasite *Plasmodium falciparum*.
  • Thiol probe for measuring unfolded protein load and proteostasis
  • Identifies proteostasis imbalances in Huntington disease models
  • Detects protein damage in malaria parasite research
  • Excitation wavelength: 350 nm
  • Emission wavelength: 470 nm
Catalog No. 50-003-55846