Learn More
Medchemexpress LLC 1H-Pyrrole-2-carboxamide, N-[(1S)-2-amino-1-[4-(hydroxymethyl)-2-thiazolyl]ethyl]-5-[3-fluoro-4-(trifluoromethyl)phenyl]- | 2234273-72-2 | 99.8% | 428.40 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13998510MM/1ML
Encompass_Preferred
NBD-14189 is a potent HIV-1 entry antagonist with an IC50 of 89 nM against the HIV-1HXB2 pseudovirus. It binds to HIV-1 gp120 and demonstrates potent antiviral activity (EC50 < 200 nM). It is intended for research use only.
  • Potent HIV-1 entry antagonist
  • Binds to HIV-1 gp120
  • Shows potent antiviral activity (EC50<200 nM)
  • Anti-HIV-1 activity (IC50=0.089 μM) in single-cycle (TZM-bl cells) assays
  • Prevents HIV-1 mediated cell-cell fusion
Catalog No. 50-003-49451