Learn More
Medchemexpress LLC 1H-Thieno[3,4-d]imidazole-4-pentanamide, N-(23-amino-3,6,9,12,15,18,21-heptaoxatricos-1-yl) hexahydro-2-oxo- | 1334172-76-7 | 98.0% | C26H50N4O9S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY140898100MG
Encompass_Preferred
Biotin-PEG7-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. These molecules contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains ligands for E3 ubiquitin ligase and target proteins
  • Exploits intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55072