Learn More
Cayman Chemical 1H1ethyl Cndesartn CIlexe 10mg
SDP
Supplier:  Cayman Chemical 1791210 MG
Encompass_Preferred
A potential impurity found in bulk preparations of candesartan cilexetil; a degradation product formed by thermal, photolytic, oxidative, or hydrolytic stress during storage
Catalog No. 50-296-5882