Learn More
AJINOMOTO USA INC L-Proline Poly Drum 1Kg
Supplier:  AJINOMOTO USA INC 30427
Encompass
Chemical Name (S)-2-Pyrrolidinecarboxylic Acid; Chemical Formula C5H9NO2; Molecular Weight; 115.13;Nitrogen Content 12.17%; CAS Number 147-85-3; Assay 99.0% to 101.0%; Description White crystals or crystalline powder; slightly sweet taste 1 Kg Poly Drum
Catalog No. NC1373112