Learn More
Ace Glass, Inc. Beaker, 1L, jacketed, Ace-Safe hose connections for 3/8intubing, 175mm inside height, 91mm inside diameter
SDP
Supplier:  Ace Glass, Inc. 5340118
Encompass_Preferred
1L jacketed beaker with one upper and one lower Ace-Safe hose connection on opposite sides, for 3/8 in. tubing. Dimensions: 175mm I.H. x 91mm I.D. Does not have a pouring spout.
Catalog No. 50-971-850