Learn More
Chemglass Life Sciences Flask, Round Bottom, 1000mL, Heavy Wall, 4-Neck, 29/26 CN - 29/26 SN's, 10/30 Thermometer Joint
SDP
Supplier:  Chemglass Life Sciences CG153307
Encompass_Preferred
Flask, Round Bottom, 1000mL, Heavy Wall, 29/26 - 29/26, 10/30 Thermometer Joint, 4-Neck. Round bottom, HEAVY WALL, four-neck flask with standard taper outer joints on the center neck and vertical side necks. The third neck is a 10/30 outer joint for use with a CG-3506, 10/30 jointed thermometer. Flasks with 29/26 center necks have a groove tooled just below the grind to aid in the clamping of the flask.
Catalog No. CG153307