Learn More
Teknova 1M MES, pH 6.0. 1000mL, Endotoxin Tested, Sterile.
SDP
Supplier:  Teknova M7410
Encompass
MES solution is a zwitterionic buffer used to help maintain pH levels in biochemistry experiments. It has a buffering range of pH 5.8 - 6.5.
Catalog No. 50-238-7971