Learn More
Teknova 1M Sodium Phosphate Buffer, pH 7.2. 2L, Sterile, DNase/RNase Tested.
SDP
Supplier:  Teknova P2072
Encompass
Sodium phosphate is a versatile chemical compound that has a wide range of uses. In molecular biology, it can be used to extract DNA and RNA from cells. In protein chemistry, it can be used to purify proteins. And in biochemical applications, it can be used to study enzyme activity. Sodium phosphate is also an effective buffering agent, making it an ideal solution for use in laboratory and industrial settings. The buffer range is 5.8 to 8.0.
Catalog No. NC0318368