Learn More
Cayman Chemical 1MethylnIcotInamIDCLOrId 500mg
SDP
Supplier:  Cayman Chemical 16604500 MG
Encompass_Preferred
An active metabolite of nicotinamide; induces nitric oxide secretion from HUVECs in a concentration-dependent manner; increases plasma levels of 6-keto-prostaglandin F1α in normotensive rats at 30 mg/kg and inhibits collagen-induced thrombus formation in rats at the same dose
Catalog No. 50-296-7390