Learn More
Thomas Scientific GelGreen Nucleic Acid Gel Stain, 10,000X in Water, 0.1mL
Supplier: Thomas Scientific C755G23
For new users, we recommend GelGreen® 10,000X in water, our latest formulation that eliminates the hazards of handling DMSO for better safety. Replaces SYBR or GelStar. Excites at 488nm.