Learn More
Leica Biosystems 1ML NCL-L-CD11C-563
Supplier:  Leica Biosystems CD11C563LCE
Encompass
Antigen Background CD11c is a member of the leukocyte integrin family of adhesion proteins. It is reported to be expressed in normal tissues, mainly on myeloid cells, for example, in bone marrow myelocytes, premyelocytes, metamyelocytes, non-segmented and segmented neutrophils with high levels reported on tissue macrophages and monocytes and with lowest levels in granulocytes. It is also reported to be expressed on NK cells, activated T cells, lymphoid cell lines, including hairy cell leukemias and a proportion of interdigitating dendritic cells. Disclaimer CD11c is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. NC1445643