Learn More
Leica Biosystems 1ml NCL-MEROSIN
Supplier:  Leica Biosystems MEROSINA
Encompass
Antigen BackgroundThe muscle-specific form of laminin, merosin, is composed of three chains: alpha 2, beta 1 and gamma 1.Mutations in the chromosome 6 encoded gene for the laminin alpha 2 chain of merosin are responsible for a form of congenital muscular dystrophy (CMD). Merosin-negative CMD is characterized by a severe clinical phenotype and is associated with white matter changes on brain imaging.DisclaimerMerosin Laminin Alpha 2 Chain is recommended for the detection of specific antigens of interest in normal and neoplastic tissues, as an adjunct to conventional histopathology using non-immunologic histochemical stains.
Catalog No. 50-255-2214